3,4,5-Trifluoroiodobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 1,2,3-Trifluoro-5-iodobenzene

012627-1G 012627-5G 012627-25G
101803-006EA 29.23 USD
101803-006 101803-008 101803-010
3,4,5-Trifluoroiodobenzene ≥98%
3,4,5-Trifluoroiodobenzene

Hazard: Irritant/Light Sensitive/Refrigerate

Formula: C₆H₂F₃I
MW: 257.9800 g/mol
MDL Number: MFCD03265259
CAS Number: 170112-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR