4-Chloro-1H-pyrazole-3-carboxaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-1H-pyrazole-5-carbaldehyde, 4-Chloro-3-formylpyrazole

012591-250MG 012591-1G
101802-932EA 127.69 USD
101802-932 101802-934
4-Chloro-1H-pyrazole-3-carboxaldehyde ≥98%
4-Chloro-1H-pyrazole-3-carboxaldehyde

Hazard: Irritant

Formula: C₄H₃ClN₂O
MW: 130.53 g/mol
MDL Number: MFCD00110641
CAS Number: 623570-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR