tert-Butoxy acetic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(tert-Butoxy)acetic acid
012582-1G
012582-5G
101802-912EA
21.54
USD
101802-912
101802-914
tert-Butoxy acetic acid ≥97%
tert-Butoxy acetic acid
Hazard: Irritant
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 104…106 °C (12 Torr) Melting Pt: –5 °C |
MDL Number:
MFCD03452738 CAS Number: 13211-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...