N-Piperazineacetic acid morpholine amide ≥90%

Supplier: Matrix Scientific

Synonyms: 1-(Morpholin-4-yl)-2-(piperazin-1-yl)ethan-1-one

012569-1G 012569-5G
101802-880EA 89.23 USD
101802-880 101802-882
N-Piperazineacetic acid morpholine amide ≥90%
N-Piperazineacetic acid morpholine amide

Hazard: Irritant

Formula: C₁₀H₁₉N₃O₂
MW: 213.28 g/mol
MDL Number: MFCD00006172
CAS Number: 39890-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR