3,6-Dichloropicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3,6-Dichloropyridine-2-carboxylic acid

012517-25G 012517-100G
101802-734EA 29.23 USD
101802-734 101802-736
3,6-Dichloropicolinic acid ≥95%
3,6-Dichloropicolinic acid

TCA listed: T
Hazard: Irritant

Formula: C₆H₃Cl₂NO₂
MW: 192.00
Melting Pt: 151-152°C
MDL Number: MFCD00078655
CAS Number: 1702-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR