Benzyl trifluoromethyl sulfide ≥98%

Supplier: Matrix Scientific

012512-5G 012512-25G
101802-720EA 80 USD
101802-720 101802-722
Benzyl trifluoromethyl sulfide ≥98%
Benzyl trifluoromethyl sulfide

Hazard: Irritant

Formula: C₈H₇F₃S
Boiling Pt: 76…77 °C (30 Torr)
MDL Number: MFCD06245933
CAS Number: 351-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR