1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Trimetazidine Dihydrochloride
012231-5G
012231-25G
101802-170EA
70.77
USD
101802-170
101802-172
1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride ≥95%
1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂N₂O₃ MW: 339.26 g/mol Melting Pt: 225…228 °C |
MDL Number:
MFCD00243086 CAS Number: 13171-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...