2,4-Dimethyl-5-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dimethylthiazole-5-carboxylic acid

008155-1G 008155-5G 008155-25G
101794-124EA 20 USD
101794-124 101794-126 101794-128
2,4-Dimethyl-5-thiazolecarboxylic acid ≥95%
2,4-Dimethyl-5-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
Melting Pt: 124°C
MDL Number: MFCD00052943
CAS Number: 53137-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR