Thiophene-2,5-dicarboxylic acid monomethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Methoxycarbonyl)thiophene-2-carboxylic acid

008150-1G 008150-5G
101794-114EA 84.62 USD
101794-114 101794-116
Thiophene-2,5-dicarboxylic acid monomethyl ester ≥95%
Thiophene-2,5-dicarboxylic acid monomethyl ester

Hazard: Irritant

Formula: C₇H₆O₄S
MW: 186.19 g/mol
Melting Pt: 187…190 °C
MDL Number: MFCD01203161
CAS Number: 50340-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR