2-Methyl-1H-benzoimidazole-5-carboxylic acid

Supplier: Matrix Scientific

007922-1G 007922-5G
101793-664EA 36.92 USD
101793-664 101793-666
2-Methyl-1H-benzoimidazole-5-carboxylic acid
2-Methyl-1H-benzoimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
Melting Pt: 301…302 °C
MDL Number: MFCD00800488
CAS Number: 709-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR