6-Quinoxalinecarboxylic acid
Supplier: Matrix Scientific
Synonyms:
Quinoxaline-6-carboxylic acid
007847-1G
007847-5G
101793-514EA
35.38
USD
101793-514
101793-516
6-Quinoxalinecarboxylic acid
6-Quinoxalinecarboxylic acid
Hazard: Irritant
Formula:
C₉H₆N₂O₂ MW: 174.16 g/mol |
MDL Number:
MFCD01365834 CAS Number: 6925-00-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...