3-Fluoro-4-methoxyphenylboronic acid

Supplier: Matrix Scientific

Synonyms: (3-Fluoro-4-methoxyphenyl)boronic acid, 3-Fluoro-4-methoxybenzeneboronic acid

007687-5G 007687-25G
101793-236EA 69.23 USD
101793-236 101793-238
3-Fluoro-4-methoxyphenylboronic acid
3-Fluoro-4-methoxyphenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Melting Pt: 192…198 °C
MDL Number: MFCD00807404
CAS Number: 149507-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR