2,5-Bis(hexyl)-1,4-dibromobenzene

Supplier: Matrix Scientific

Synonyms: 1,4-Dibromo-2,5-dihexylbenzene

007670-1G 007670-5G
101793-206EA 103.08 USD
101793-206 101793-208
2,5-Bis(hexyl)-1,4-dibromobenzene
2,5-Bis(hexyl)-1,4-dibromobenzene

Hazard: Irritant

Formula: C₁₈H₂₈Br₂
MW: 404.24 g/mol
MDL Number: MFCD01318993
CAS Number: 117635-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR