3,3-Difluoropiperidine hydrochloride ≥99%

Supplier: Matrix Scientific

007653-250MG 007653-1G
101793-186EA 115.38 USD
101793-186 101793-188
3,3-Difluoropiperidine hydrochloride ≥99%
3,3-Difluoropiperidine hydrochloride

Hazard: Irritant/Hygroscopic

Formula: C₅H₁₀ClF₂N
MW: 157.59 g/mol
Melting Pt: 236…239 °C (dec.)
MDL Number: MFCD03452800
CAS Number: 496807-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR