2-(N-Methyl-N-butylamino)ethyl chloride hydrochloride ≥97%

Supplier: Matrix Scientific

007620-5G 007620-25G
101793-128EA 52.31 USD
101793-128 101793-130
2-(N-Methyl-N-butylamino)ethyl chloride hydrochloride ≥97%
2-(N-Methyl-N-butylamino)ethyl chloride hydrochloride

Hazard: Irritant

Formula: C₇H₁₇Cl₂N
Melting Pt: 119…120 °C
MDL Number: MFCD00050521
CAS Number: 115571-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR