10-Bromodecanoic acid ≥95%

Supplier: Matrix Scientific

007516-25G 007516-100G
101792-962EA 60 USD
101792-962 101792-964
10-Bromodecanoic acid ≥95%
10-Bromodecanoic acid

Hazard: Irritant

Formula: C₁₀H₁₉BrO₂
Boiling Pt: 160 °C (1 Torr)
Melting Pt: 39…40 °C
MDL Number: MFCD00014388
CAS Number: 50530-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR