5-Phenyl-o-phenetidine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-ethoxybiphenyl

007487-1G 007487-5G
101792-908EA 158.46 USD
101792-908 101792-910
5-Phenyl-o-phenetidine ≥98%
5-Phenyl-o-phenetidine

Hazard: Irritant

Formula: C₁₄H₁₅NO
MW: 213.28 g/mol
MDL Number: MFCD03425697
CAS Number: 856343-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR