3-(Trifluoromethyl)biphenyl ≥98%

Supplier: Matrix Scientific

007484-1G 007484-5G
101792-904EA 141.54 USD
101792-904 101792-906
3-(Trifluoromethyl)biphenyl ≥98%
3-(Trifluoromethyl)biphenyl

Hazard: Irritant

Formula: C₁₃H₉F₃
Boiling Pt: 117-118°C/13 Torr
Melting Pt: 26…27 °C
MDL Number: MFCD03426089
CAS Number: 366-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR