4-Methoxy-2-methylphenylacetonitrile ≥97%

Supplier: Matrix Scientific

007476-1G 007476-5G
101792-882EA 107.69 USD
101792-882 101792-884
4-Methoxy-2-methylphenylacetonitrile ≥97%
4-Methoxy-2-methylphenylacetonitrile

Hazard: Irritant

Formula: C₁₀H₁₁NO
MDL Number: MFCD02178388
CAS Number: 262298-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR