2-(Heptafluoro-n-propyl)benzimidazole

Supplier: Matrix Scientific

007449-1G 007449-5G
101792-790EA 72.31 USD
101792-790 101792-792
2-(Heptafluoro-n-propyl)benzimidazole
2-(Heptafluoro-n-propyl)benzimidazole

Hazard: Irritant

Formula: C₁₀H₅F₇N₂
MW: 286.15 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00521800
CAS Number: 559-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR