4-Fluoropiperidine hydrochloride ≥98%

Supplier: Matrix Scientific

007437-1G 007437-5G
101792-768EA 76.92 USD
101792-768 101792-770
4-Fluoropiperidine hydrochloride ≥98%
4-Fluoropiperidine hydrochloride

Hazard: Irritant/Air Sensitive

Formula: C₅H₁₁ClFN
Melting Pt: 163…164 °C
MDL Number: MFCD03452786
CAS Number: 57395-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR