2-Phenylethylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Phenethylboronic acid

007417-5G 007417-25G
101792-702EA 63.08 USD
101792-702 101792-704
2-Phenylethylboronic acid ≥97%
2-Phenylethylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD01631226
CAS Number: 34420-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR