3-Fluoro-4-methylphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-methylbenzeneboronic acid

007408-1G 007408-5G
101792-682EA 56.92 USD
101792-682 101792-684
3-Fluoro-4-methylphenylboronic acid ≥97%
3-Fluoro-4-methylphenylboronic acid

Hazard: Irritant

Formula: C₇H₈BFO₂
Melting Pt: 234 °C
MDL Number: MFCD02683115
CAS Number: 168267-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR