4-Iodo-2-methylanisole ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Iodo-1-methoxy-2-methylbenzene
007383-5G
007383-25G
101792-610EA
80
USD
101792-610
101792-612
4-Iodo-2-methylanisole ≥97%
4-Iodo-2-methylanisole
Hazard: Irritant
Formula:
C₈H₉IO MW: 248.06 g/mol Melting Pt: 75…76 °C |
MDL Number:
MFCD03425972 CAS Number: 75581-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...