(3-Carbamoylphenyl)boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (3-carbamoylphenyl)boronic acid, 3-Aminocarbonylphenylboronic acid, 3-carbamoylbenzeneboronic acid, 3-Carbamoylphenylboronic acid, 3-Carbamoylbenzeneboronic acid

007364-1G 007364-5G
101792-558EA 15.38 USD
101792-558 101792-560
(3-Carbamoylphenyl)boronic acid ≥97%
(3-Carbamoylphenyl)boronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₈BNO₃
MW: 164.955 g/mol
Melting Pt: 232…238 °C
MDL Number: MFCD03411948
CAS Number: 351422-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR