1,1-Dichloro-3,3,3-trifluoropropene ≥99%

Supplier: Matrix Scientific

007200-5G 007200-25G
101792-178EA 144.62 USD
101792-178 101792-180
1,1-Dichloro-3,3,3-trifluoropropene ≥99%
1,1-Dichloro-3,3,3-trifluoropropene

TCA listed: T
Hazard: Irritant

Formula: C₃HCl₂F₃
MW: 164.94 g/mol
Boiling Pt: 54 °C
Density: 1.4605
MDL Number: MFCD02184272
CAS Number: 460-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR