Ethyl-4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

007024-1G 007024-5G
101791-854EA 75.38 USD
101791-854 101791-856
Ethyl-4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate ≥97%
Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₃
MW: 285.2186 g/mol
Melting Pt: 216 °C
MDL Number: MFCD09878378
CAS Number: 23851-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR