Ethyl-4-hydroxy-7-(trifluoromethyl)quinoline-3-carboxylate

Supplier: Matrix Scientific

007023-1G 007023-5G
101791-850EA 124.62 USD
101791-850 101791-852
Ethyl-4-hydroxy-7-(trifluoromethyl)quinoline-3-carboxylate
Ethyl 4-hydroxy-7-(trifluoromethyl)quinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₃
MW: 285.22 g/mol
Melting Pt: 294…297 °C
MDL Number: MFCD00006769
CAS Number: 391-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR