Ethyl-4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

007022-1G 007022-5G
101791-846EA 75.38 USD
101791-846 101791-848
Ethyl-4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate
Ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₃
MW: 285.2200 g/mol
MDL Number: MFCD00173369
CAS Number: 26893-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR