3-(4-Chlorophenyl)pyrazole

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)-1H-pyrazole

007005-1G 007005-5G
101791-800EA 16.92 USD
101791-800 101791-802
3-(4-Chlorophenyl)pyrazole
3-(4-Chlorophenyl)pyrazole

Hazard: Toxic

Formula: C₉H₇ClN₂
MW: 178.62 g/mol
Melting Pt: 100…104 °C
MDL Number: MFCD00111881
CAS Number: 59843-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR