2-Chloro-N-(2-fluorophenyl)acetamide ≥97%

Supplier: Matrix Scientific

007001-1G 007001-5G
101791-790EA 95.38 USD
101791-790 101791-792
2-Chloro-N-(2-fluorophenyl)acetamide ≥97%
2-Chloro-N-(2-fluorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₇ClFNO
Melting Pt: 88…90 °C
MDL Number: MFCD00730970
CAS Number: 347-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR