3-Quinolineboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Quinoline-3-boronic acid, Quinolin-3-ylboronic acid

006792-1G 006792-5G
101791-128EA 24.62 USD
101791-128 101791-130
3-Quinolineboronic acid ≥97%
3-Quinolineboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₉H₈BNO₂
Melting Pt: >300 °C
MDL Number: MFCD02183527
CAS Number: 191162-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR