2-Methoxy-5-pyridineboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (6-Methoxypyridin-3-yl)boronic acid

006786-1G 006786-5G
101791-108EA 15.38 USD
101791-108 101791-110
2-Methoxy-5-pyridineboronic acid ≥95%
2-Methoxy-5-pyridineboronic acid

Hazard: Irritant/Air Sensitive/Store in Freezer

Formula: C₆H₈BNO₃
MW: 152.95 g/mol
Melting Pt: 132°C
MDL Number: MFCD02093044
CAS Number: 163105-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR