3-(Isopropoxycarbonyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Isopropoxycarbonyl)benzeneboronic acid

006782-1G 006782-5G
101791-102EA 95.38 USD
101791-102 102550-888
3-(Isopropoxycarbonyl)phenylboronic acid ≥97%
3-(Isopropoxycarbonyl)phenylboronic acid

Hazard: Irritant

Formula: C₁₀H₁₃BO₄
MW: 208.02
Melting Pt: 127…129 °C
MDL Number: MFCD02093047
CAS Number: 342002-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR