3-(Ethoxycarbonyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

006777-1G 006777-5G 006777-25G
101791-084EA 15.38 USD
101791-084 101791-086 101791-088
3-(Ethoxycarbonyl)phenylboronic acid ≥97%
3-(Ethoxycarbonyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₉H₁₁BO₄
Melting Pt: 135-139°C
MDL Number: MFCD02179444
CAS Number: 4334-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR