Copper(I)-2-thiophenecarboxylate ≥96%

Supplier: Matrix Scientific

Synonyms: (2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC

006771-5G 006771-25G
101791-064EA 44.62 USD
101791-064 101791-066
Copper(I)-2-thiophenecarboxylate ≥96%
Copper(I)-2-thiophenecarboxylate

Hazard: Irritant/Light Sensitive/Air Sensitive/Refrigerate

Formula: C₅H₃CuO₂S
MW: 190.68 g/mol
Boiling Pt: 260 °C
Flash Pt: 117 °C
MDL Number: MFCD02183524
CAS Number: 68986-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR