4-(Aminomethyl)phenylboronic acid hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Aminomethylphenylboronic acid hydrochloride, 4-(Aminomethyl)benzeneboronic acid hydrochloride

006751-1G 006751-5G
101791-008EA 80 USD
101791-008 101791-010
4-(Aminomethyl)phenylboronic acid hydrochloride ≥97%
4-(Aminomethyl)phenylboronic acid hydrochloride

Hazard: Irritant/Refrigerate

Formula: C₇H₁₁BClNO₂
MW: 187.43 g/mol
Melting Pt: 240…250 °C
MDL Number: MFCD01632199
CAS Number: 75705-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR