5-Formyl-2-furanylboronic acid ≥98%

Supplier: Matrix Scientific

006509-1G 006509-25G
101790-666EA 15.38 USD
101790-666 101790-670
5-Formyl-2-furanylboronic acid ≥98%
5-Formyl-2-furanylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₅H₅BO₄
Melting Pt: 150…151 °C
MDL Number: MFCD01114696
CAS Number: 27329-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR