3,4'-Bis(trifluoromethyl)benzhydrol ≥97%
Supplier: Matrix Scientific
006372-1G
006372-5G
101790-436EA
55.38
USD
101790-436
101790-438
3,4'-Bis(trifluoromethyl)benzhydrol ≥97%
3,4'-Bis(trifluoromethyl)benzhydrol
Hazard: Irritant
Formula:
C₁₅H₁₀F₆O MW: 320.23 g/mol |
MDL Number:
MFCD00045091 CAS Number: 203915-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...