Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate ≥90%

Supplier: Matrix Scientific

006192-5G 006192-25G 006192-100G
101790-228EA 63.08 USD
101790-228 101790-230 101790-232
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate ≥90%
Trimethylsilyl 2,2-difluoro-2-(fluorosulphonyl)acetate

Hazard: Flammable/Corrosive

Formula: C₅H₉F₃O₄SSi
MW: 250.27 g/mol
Boiling Pt: 156°C
Density: 1.27
Flash Pt: 26°C
MDL Number: MFCD02093343
CAS Number: 120801-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR