1-(2-Fluorophenyl)piperazine hydrochloride

Supplier: Matrix Scientific

006106-5G 006106-25G
101790-088EA 90.77 USD
101790-088 101790-090
1-(2-Fluorophenyl)piperazine hydrochloride
1-(2-Fluorophenyl)piperazine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃FN₂
Melting Pt: 185…186 °C
MDL Number: MFCD00134409
CAS Number: 1011-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR