1-(2-Fluorophenyl)biguanide hydrochloride ≥97%

Supplier: Matrix Scientific

006084-1G 006084-5G
101790-032EA 69.23 USD
101790-032 101790-034
1-(2-Fluorophenyl)biguanide hydrochloride ≥97%
1-(2-Fluorophenyl)biguanide hydrochloride

Hazard: Irritant

Formula: C₈H₁₁ClFN₅
MDL Number: MFCD00236682
CAS Number: 66088-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR