2-(Difluoromethylthio)benzamide

Supplier: Matrix Scientific

006031-1G 006031-5G
101789-956EA 36.92 USD
101789-956 101789-958
2-(Difluoromethylthio)benzamide
2-(Difluoromethylthio)benzamide

Hazard: Irritant

Formula: C₈H₇F₂NOS
CAS Number: 261944-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR