4-(Difluoromethoxy)benzylamine

Supplier: Matrix Scientific

Synonyms: (4-(Difluoromethoxy)phenyl)methanamine

006007-1G 006007-5G
101789-906EA 36.92 USD
101789-906 101789-908
4-(Difluoromethoxy)benzylamine
4-(Difluoromethoxy)benzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₈H₉F₂NO
MDL Number: MFCD00236231
CAS Number: 177842-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR