2,4-Bis(trifluoromethyl)phenylacetonitrile
Supplier: Matrix Scientific
005841-1G
005841-5G
101789-628EA
95.38
USD
101789-628
101789-630
2,4-Bis(trifluoromethyl)phenylacetonitrile
2,4-Bis(trifluoromethyl)phenylacetonitrile
Hazard: Irritant
Formula:
C₁₀H₅F₆N |
CAS Number:
201789-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...