1-Bromo-2,6-bis(trifluoromethyl)benzene ≥98%

Supplier: Matrix Scientific

005839-1G 005839-5G 005839-25G
101789-618EA 50.77 USD
101789-618 101789-620 101789-622
1-Bromo-2,6-bis(trifluoromethyl)benzene ≥98%
1-Bromo-2,6-bis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₃BrF₆
Boiling Pt: 86°C/29mmHg
Melting Pt: 51-52°C
MDL Number: MFCD00236210
CAS Number: 118527-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR