5,7-Bis(trifluoromethyl)-4-hydroxyquinoline ≥97%

Supplier: Matrix Scientific

005836-250MG 005836-1G
101789-614EA 63.08 USD
101789-614 101789-616
5,7-Bis(trifluoromethyl)-4-hydroxyquinoline ≥97%
5,7-Bis(trifluoromethyl)-4-hydroxyquinoline

Hazard: Irritant

Formula: C₁₁H₅F₆NO
MDL Number: MFCD00153141
CAS Number: 237076-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR