4-Dibenzofuranboronic acid

Supplier: Matrix Scientific

Synonyms: Dibenzofuran-4-boronic acid, Dibenzo[b,d]furan-4-ylboronic acid

005767-1G 005767-5G
101789-484EA 23.08 USD
101789-484 101789-486
4-Dibenzofuranboronic acid
4-Dibenzofuranboronic acid

Hazard: Irritant/Refrigerate

Formula: C₁₂H₉BO₃
MW: 212.01 g/mol
Melting Pt: 286…291 °C
MDL Number: MFCD00092336
CAS Number: 100124-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR