3-Carboxyphenylboronic acid

Supplier: Matrix Scientific

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

005715-5G 005715-25G
101789-416EA 16.92 USD
101789-416 101789-418
3-Carboxyphenylboronic acid
3-Carboxyphenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₇BO₄
MW: 165.94 g/mol
Melting Pt: 245-247°C
MDL Number: MFCD00036833
CAS Number: 25487-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR