2-(Perfluorooctyl)ethyl allyl sulfide

Supplier: Matrix Scientific

005553-1G 005553-5G
101789-114EA 76.92 USD
101789-114 101789-116
2-(Perfluorooctyl)ethyl allyl sulfide
2-(Perfluorooctyl)ethyl allyl sulfide

Hazard: Irritant

Formula: C₁₃H₉F₁₇S
MDL Number: MFCD01862012
CAS Number: 239463-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR